missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10355710

Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10355710
1 L, HDPE plastic bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10355710

Brand: Fisher Chemical J/8460/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oilShow MoreShow Less
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg at 20°C
Quantity 1 L
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2526689 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberJ/8460/25,J/8460/24,J/8460C/90,J/8460C/05,J/8460/17,J/8460/15
Product Identifier
  • Sulfuric acid, 0.1N (0.05M) standard solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.