missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid Solution 0.1M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10723611

Sulfuric Acid Solution 0.1M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10723611
1 L, HDPE plastic bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy

Product Code. 10723611

Brand: Fisher Chemical J/8450/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sulfuric Acid Solution 0.1M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oilShow MoreShow Less
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg at 20°C
Quantity 1 L
Physical Form Liquid
Product Identifier
  • Sulfuric acid, 0.1N (0.05M) standard solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid Solution 0.1M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.