missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.1M, Reag. Ph. Eur., Honeywell Fluka™

Product Code. 15694990
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Volumetric, Reag. Ph. Eur., 0.1 M H2SO4 (0.2N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Concentration 0.1M
Quantity 1 L
Linear Formula H2SO4
UN Number UN3264
Beilstein 2037554
Formula Weight 98.08g/mol
Grade Reagent Ph. Eur.
Chemical Name or Material Sulfuric acid solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'