missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid solution, 0.25M, Reag. Ph. Eur., Honeywell Fluka™
45.83 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
10 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 0.25M |
Packaging | Plastic Bottle |
Quantity | 1 L |
Linear Formula | H2SO4 |
UN Number | UN3264 |
Beilstein | 2037554 |
Formula Weight | 98.08g/mol |
Grade | Reagent Ph. Eur. |
Chemical Name or Material | Sulfuric acid solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'