missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™
54.50 € - 166.00 €
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10418030
|
Fisher Chemical
J/8430/17 |
2.5 L | HDPE plastic bottle |
54.50 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12688635
|
Fisher Chemical
J/8430/25 |
25 L | Plastic container | N/A | |||||
Description
Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
1.5 | |
7664-93-9 | |
1mmHg at 20°C | |
Liquid | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Colorless | |
100°C | |
H2O4S | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sulfuric acid, solution 0.5M, (1N)
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.