missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations

Specifications
Specifications
Concentration | 0.5M |
Packaging | Plastic Bottle |
Quantity | 2.5 L |
Linear Formula | H2SO4 |
UN Number | UN3264 |
Beilstein | 2037554 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Formula Weight | 98.08g/mol |
Grade | Reagent Ph. Eur. |
Chemical Name or Material | Sulfuric acid solution |
Safety and Handling

Product Identifier
- Sulfuric acid solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction