missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution 1M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 11498962
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy

Sulfuric Acid Solution 1M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1
Color Colorless
Boiling Point 100°C
Packaging Bag in box
Vapor Pressure 1mmHg at 20°C
Quantity 10 L
Physical Form Liquid
Formula Weight 98.07g/mol
compliance-icons compliance-icons
Product Identifier
  • Sulfuric acid M
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'