missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, TraceSELECT™ Ultra, ≥95% (T), Honeywell Fluka™

Product Code. 15617060
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Poly bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

TraceSELECT™ Ultra, for ultratrace analysis,≥95%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH <1
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cm3 at 20°C
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (146°C)
Quantity 500 mL
Linear Formula H2SO4
Physical Form Liquid
UN Number UN1830
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity ≥95% (T)
Grade TraceSELECT™ Ultra
Chemical Name or Material Sulfuric acid
Show More Show Less
compliance-icons
Product Identifier
  • sulphuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'