missing translation for 'onlineSavingsMsg'
Learn More

Sulfurous Acid, Pure, Approx.6% (w/v) SO2, Fisher Chemical

Product Code. 10395800
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle

Specifications

Color Colorless
CAS 7782-99-2
Packaging Amber glass bottle
Quantity 2.5 L
Molecular Formula H2O3S
Physical Form Liquid
MDL Number 11335
Synonym sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
SMILES OS(=O)O
IUPAC Name sulfurous acid
Molecular Weight (g/mol) 82.073
PubChem CID 1100
ChEBI CHEBI:48854
Formula Weight 82.0722g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfurous Acid, pure, approx. 6% w/v SO5
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.