Learn More
Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 378158000
217.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Sulfuryl chloride | |
7791-25-5, 75-09-2 | |
Cl2O2S | |
MFCD00011451 | |
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride | |
ClS(Cl)(=O)=O | |
134.96 | |
CHEBI:29291 | |
1M, exact strength on the certificate of analysis | |
Clear pale yellow solution | |
1.3520g/mL |
1.0M Solution in Dichloromethane | |
800 mL | |
SO2Cl2 | |
15, 9110 | |
YBBRCQOCSYXUOC-UHFFFAOYSA-N | |
sulfuryl dichloride | |
24648 | |
134.97 | |
AcroSeal™ Glass Bottle | |
1.352 |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Sulfuryl chloride, 1M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H331-Toxic if inhaled.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467154 | Certificate of Analysis | 30/01/2025 | 378158000,378151000 |
A0462944 | Certificate of Analysis | 17/06/2024 | 378158000,378151000 |
A0453217 | Certificate of Analysis | 25/08/2023 | 378158000,378151000 |
A0413955 | Certificate of Analysis | 13/12/2022 | 378158000,378151000 |
A0401306 | Certificate of Analysis | 12/12/2022 | 378158000,378151000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only