missing translation for 'onlineSavingsMsg'
Learn More
Sulfuric Acid, Dilute, EP Grade, Reagecon™ Product Code.: 11414749
compliance-icons compliance-icons

Sulfuric Acid, Dilute, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11414749 Shop All Reagecon Products
1 L, Tamper evident
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
This item is not returnable. View return policy

Product Code. 11414749

Brand: Reagecon 1086804

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Sulfuric Acid, Dilute, EP Grade, Reagecon™ is a ready-to-use reagent with 98g/L of H2SO4 in water for use in manual and automated titrations.

Dilute sulfuric acid is a strong acid and a good electrolyte. It is highly ionized and turns blue litmus red. It reacts with many metals, most hydroxides and oxides, some carbonates and sulfides, and some salts. This solution is prepared by adding 5.5mL sulphuric acid R to 60mL water, cooled and then diluted to 100mL, for a final concentration of 1M (98g/L).
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions

Specifications

CAS 7664-93-9, 7732-18-5
Composition 5.5mL sulfuric acid in 60mL water, diluted to 100mL with water
Concentration 1M
Odor Odorless
Packaging Tamper evident
Certificate of Analysis Yes
Solvent or Matrix Water
Quantity 1 L
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.1g/mol
Concentration or Composition (by Analyte or Components) 98g/L H2SO4
Chemical Name or Material Sulfuric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSU2124G1 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number1086804
Lot NumberSU2123G1 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number1086804
Lot NumberSU2122K1 Certificate TypeCertificate of Analysis Date03/05/2023 Catalog Number1086804
Lot NumberSU2121C1 Certificate TypeCertificate of Analysis Date20/04/2022 Catalog Number1086804
compliance-icons compliance-icons
Product Identifier
  • SULPHURIC ACID, DILUTE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sulfuric Acid, Dilute, EP Grade, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.