missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid solution 0.005M, Fisher Chemical™

Product Code. 12940774
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 7664-93-9
Concentration 0.005M (0.01N)
Packaging Bottle
Quantity 100 mL
Molecular Formula H2O4S
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Chemical Name or Material Sulphuric Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'