missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulphuric Acid Solution, 0.05M, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Specifications
Specifications
Concentration | 0.05, 0.1mol/L, eq/L |
Quantity | 1000 mL |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'