missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ Suma Alcohol Wipes
No-rinse alcoholic disinfectant wipes
133.00 €
Specifications
Length (Metric) | 135 mm |
---|---|
Width (Metric) | 180 mm |
Dimensions (L x W) | 135 x 180 mm |
Packaging Quantity | 12 x 150 Wipes |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17603095
|
Diversey™
100878518 |
12 x 150 Wipes for AT, DE, FR, GB, IE only | |||||||
Description
Suma Alcohol Wipes are ready-to-use wipes for disinfection of all hard surfaces like work areas, small equipment, utensils, and handholds.
Key properties
- Suma Alcohol Wipes are perfect for disinfection of all food contact areas and can be used as complement to your cleaning and disinfection procedures.
- The product is based on ethanol to provide fast and effective disinfection.
- It is suitable for the disinfection of alcohol-safe, food contact surfaces and articles such as chopping boards, utensils, thermometers, tables and other smaller kitchen equipment.
- It is effective against a wide spectrum of micro-organisms.
Benefits
- Easy to use
- No spraying, safe to use
- Effective against a wide range of micro-organisms
- Quick drying minimizes disruption to work flow
Specifications
135 mm | |
180 mm | |
135 x 180 mm | |
12 x 150 Wipes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X1800 Suma Alcohol Wipe, 150/pack, 12 packs - AT, DE, FR, GB & IE only
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : ethanol, propan-2-ol