missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Adjustable Super Erecta™ Wire Shelf, Chrome Finish

Unique, open-wire design of chrome finish shelf permits many configurations, free circulation of air, and minimizes dust accumulation.

Brand:  Metro™ A2124NC

 View more versions of this product

Product Code. 15906831

  • 107.00 € / Each
Estimated Shipment: 02-09-2025
to see stock.
Explore more special offers
Add to basket

Includes:

Each shelf shipped with one pack of four nonconductive sleeve/wedges.

This item is not returnable. View return policy

Description

Description

  • Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items.
  • Can be loaded and unloaded easily from all sides.
  • Hard, durable surface.
  • Abrasion resistant.
  • Ideal for dry, noncorrosive environments.
  • Shelves adust in 1 in. increments with patented shelf-release corners.
  • Shelf releases and wedges offer improved rigidity.
  • Assembles quickly without tools; assembled units are nonconductive.

Order Info

Posts sold separately.

Specifications

Specifications

Wire Shelf
Ideal for Drying, Non-corrosive Environments
Four non conductive sleeve/wedges
61 cm
53 cm
24 x 21 in. (61 x 53 cm)
Metro™ Super Erecta™ adjustable shelving system
24 in.
21 in.
Chrome
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Chrome Finish > 21 x 24 in. (53 x 61cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.