Learn More
Cytiva Superdex™ 200 3.2/300 Prepacked Precision Column
Sensitive and high-resolution gel filtration separations for biomolecules in micropreparative scale
Brand:  Cytiva 29036232
Product Code. 15127966
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Prepacked with Superdex 200 media
  • For biomolecules with molecular weights between 10,000 and 6,00,000
  • Spherical composite of cross-linked agarose and dextran
  • Supplied with two fingertight 1/16 in. male connectors for connection
  • Used for polishing step in purfication process
  • With new hardware design for direct connection to ÄAKTATM or other systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 to 0.10 mL/min.
13 μm
Superdex Pre-packed Column
Borosilicate Glass