missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superdex™ 200 Prep Grade

Produce separation medium with high selectivity and resolution. Cytiva Life Sciences™ Superdex™ 200 Prep Grade for purifying proteins of fractionation range: 10000 to 600000D

Brand:  Cytiva 17-1043-01


Product Code. 11570804

  • 845.00 € / 150mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gel filtration medium
  • For purifying proteins of fractionation range: 10000 to 600000Da
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH stability working range
  • Available in 150mL bottle

TRUSTED_SUSTAINABILITY
Specifications

Specifications

150mL
24 to 44 μm
Solid Spherical Bead
70psi
Composite of cross-linked Agarose and Dextran
4 to 30°C, 20% Ethanol
10 to 50 cm/hr.
Superdex Media
Separation, polishing and product formulation of Monoclonal Antibodies
3 to 13
Recombinant Protein
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Superdex 200 prep grade, 150 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.