missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperFi™ Buffers

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.

96.25 € - 97.50 €

Specifications

Polymerase Platinum SuperFi DNA Polymerase
Quantity 4 x 1.25 mL
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Starting Material DNA
View More Specs

Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
15555033
View Documents
Invitrogen™
12355005
Colorless
96.25 €
5mL
Please to purchase this item. Need a web account? Register with us today!
15565033
View Documents
Invitrogen™
12356005
Green
97.50 €
5mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

Specifications

Specifications

Platinum SuperFi DNA Polymerase
Dry Ice
DNA
4 x 1.25 mL
Hot-start PCR, High-fidelity PCR
4 x 1.25 mL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.