missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ BP5MS GC Capillary Column
SGE GC column BP5MS is an optimized silphenylene column for general purpose MS analysis
Brand: Trajan Scientific and Medical™ 054310
This item is not returnable.
View return policy
Description
- Perfect for 5% GCMS analysis
- Optimized silphenylene content for general purpose MS analyses
Specifications
350°C | |
-40°C to +330/350°C | |
5% Phenyl Polysilphenylene-siloxane | |
GC Capillary column |
5% GC-MS Analysis | |
30 m | |
0.25 mm | |
0.25 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction