missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ SuperFrost Plus™ Adhesion slides
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™ SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations.
Brand: Epredia™ J1820AMNZ
31.37 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- No time wasted on slide-coating procedures
- No money wasted on expensive adhesives
- Reliable and uniform coating
- Minimal sample loss
- Reliable and safe adhesive action
Specifications
75 mm | |
90°, Ground | |
Pink | |
72/Pk. |
25 mm | |
Adhesion Slide | |
Traditional |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction