Learn More
Epredia™ SuperFrost Plus™ Adhesion slides, Special Size
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™TM SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations. The printed tab ink is impervious to virtually all common laboratory chemicals and reagents. For tissue slices 2 to 5μm thick.
Brand: Epredia™ J1800BMNZ
70.59 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
- No time wasted on slide-coating procedures
- No money wasted on expensive adhesives
- Reliable and uniform coating
- Minimal sample loss
- Reliable and safe adhesive action
Specifications
75 mm | |
90°, Ground | |
White |
50 mm | |
Adhesion Slide | |
72/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.