missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ SuperFrost™ Microscope Slides, Ground 90°

Product Code. 12312108 Shop All Epredia Products
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Quantity:
50/Box, 2000 in outer carton
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 12312108

Brand: Epredia™ AG00008432E01FST20

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.

  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 90°, Ground
Thickness (Metric) 1 mm
Color Green
Certifications/Compliance ISO8037/I
Coating Uncoated
Quantity 50/Box, 2000 in outer carton
Writing Surface Yes
Dimensions (L x W) 76 x 26 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.