missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV CellsDirect™ Lysis Reagents

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples.

Brand:  Invitrogen™ 11750550

1299.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 17103674

  • 2165.00 € / 500 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The SuperScript IV CellsDirect Lysis Reagents kit includes lysis solution, stop solution, and lysis enhancer. These reagents are part of the SuperScript IV CellsDirect cDNA Synthesis Kit and are made available separately here for use when processing a large number of samples. See the SuperScript IV CellsDirect cDNA Synthesis Kit manual for protocol recommendations.

SuperScript IV CellsDirect Lysis Reagents are optimized for synthesizing first-strand cDNA directly from a mammalian cell lysate without first isolating RNA.

Features of SuperScript IV CellsDirect Lysis Reagents include:
• Ready-to-use pre-mixed cell lysis solution compatible with a variety of cell-types
• Optimized performance for 1–10,000 cells per sample; results equivalent to those from purified RNA

Note: This procedure is optimized for a maximum of 10,000 cells per reaction. If needed, volumes can be adjusted for cell numbers greater than 10,000. Using too many cells per lysis reaction can result in incomplete lysis and/or inhibition of RT-PCR.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• SuperScript IV CellsDirect Lysis Solution (9 mL)
• SuperScript IV CellsDirect Stop Solution (1.25 mL)
• Lysis Enhancer (250 μL)

Store at –20°C.

Reverse Transcription
SuperScript IV
500 reactions
Reverse Transcription
Kit
SuperScript IV CellsDirect cDNA Synthesis Kit
Dry Ice
Master Mix
Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers