missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV First-Strand Synthesis System

The SuperScript IV First-Strand Synthesis System for RT-PCR, available with or without ezDNase, is optimized for synthesis of first-strand cDNA from purified poly(A)+ or total RNA.

684.00 € - 2685.00 €

Specifications

Format Kit
GC-Rich PCR Performance High
Reaction Speed 10 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
View More Specs

Products 4
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
15327696
View Documents
Invitrogen™
18091050
Kit only 50 Reactions
684.00 €
50 reactions
Please to purchase this item. Need a web account? Register with us today!
15337696
View Documents
Invitrogen™
18091200
Kit only 200 Reactions
2465.00 €
200 reactions
Please to purchase this item. Need a web account? Register with us today!
16397225
View Documents
Invitrogen™
18091150
Kit with ezDNase 50 Reactions
738.00 €
50 reactions
Please to purchase this item. Need a web account? Register with us today!
16272082
View Documents
Invitrogen™
18091300
Kit with ezDNase 200 Reactions
2685.00 €
200 tests
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Kit
10 min.
50°C
SuperScript IV
Separate components
Up to 12.3 kb
High
Reverse Transcription
Each
First-Strand cDNA
Reverse Transcription
RNA
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SuperScript™ IV First-Strand Synthesis System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.