Learn More
Invitrogen™ SuperScript™ IV One-Step RT-PCR System
The Invitrogen SuperScript One-Step RT-PCR System, available with or without ezDNase, combines the high processivity of SuperScript IV Reverse Transcriptase (RT) with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched one-step RT-PCR performance.
Brand: Invitrogen™ 12594100
Includes:
- SuperScript IV RT Mix, 12.5µL
- 2X Platinum SuperFi RT-PCR Master Mix, 625µL
- Nuclease-free water, 1.25mL
Description
The Invitrogen SuperScript One-Step RT-PCR System, available with or without ezDNase, combines the high processivity of SuperScript IV Reverse Transcriptase (RT) with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched one-step RT-PCR performance. With this convenient one-step formulation plus gene-specific primers, you can perform both cDNA synthesis from total RNA or mRNA and PCR amplification in a single tube.
Features of the SuperScript IV One-Step RT-PCR System include:
• Superior performance with sensitivity down to 0.01 pg RNA, target length up to 13.8 kb, and the fastest one-step RT-PCR protocol
• Reliable target detection even with suboptimally pure RNA samples
• Two-phase hot-start activation mechanism for high sensitivity, specificity, and room temperature setup
• Fast and easy gDNA removal for superior accuracy and confidence in results (SuperScript IV One-Step RT-PCR System with ezDNase only)
SuperScript IV RT is a proprietary MMLV RT mutant with increased processivity, thermostability, and inhibitor resistance, as well as reduced RNase H activity, for sensitive and efficient full-length cDNA synthesis. Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR. The combination of these two engineered enzymes produces enhanced RT-PCR yields and fidelity, as well as amplification of longer templates compared to other one-step RT-PCR kits.
The inclusion of ezDNase (SuperScript IV One-Step RT-PCR System with ezDNase only) for use upstream of the RT step further enhances the one-step RT-PCR workflow through an extremely simplified genomic DNA removal step that ensures the highest accuracy and confidence in one-step RT-PCR results.
Specifications
• SuperScript IV RT Mix (50 μL) |
|
Kit | |
1-step RT-PCR | |
10 min. | |
45°C to 60°C | |
SuperScript IV | |
Dry Ice | |
Built-In Hot Start | |
Master Mix | |
Up to 14 kb | |
Kit only |
Gel Electrophoresis | |
High | |
Platinum SuperFi | |
1-step RT-PCR | |
Each | |
Reduced | |
PCR Amplified cDNA | |
100 Reactions | |
Reverse Transcription | |
RNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.