missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV RT Reaction Buffer

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase.

Brand:  Invitrogen™ 18090050B

124.60 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 15658068

  • 178.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SuperScript IV RT Reaction Buffer is included with, and designed for use with, SuperScript IV Reverse Transcriptase. This buffer is optimized to give robust and reliable cDNA synthesis. In cases where additional quantity is required, SuperScript IV RT Reaction Buffer is made available here in a stand-alone format.

For use with
SuperScript IV Reverse Transcriptase (Cat. No. 18090010)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• SuperScript IV RT Reaction Buffer (5 x 1 mL)

Store at –20°C.

SuperScript IV Reverse Transcriptase
Dry Ice
Separate components
RNA
Reverse Transcription
5 x 1 mL
5X
Reverse Transcription
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.