missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ VILO™ Master Mix

SuperScript VILO Master Mix is a first-strand cDNA synthesis master mix for two-step RT-qPCR applications.

Brand:  Invitrogen™ 11755250

 View more versions of this product

Product Code. 12023679

  • 3815.00 € / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SuperScript VILO Master Mix is a first-strand cDNA synthesis master mix for two-step RT-qPCR applications. The master mix combines SuperScript III Reverse Transcriptase (RT) and other RT reaction components in a single-tube format to enable reliable first-strand synthesis and high cDNA yields from a wide range of input RNA concentrations. The high RT efficiency of SuperScript III RT and the optimized formulation allow the master mix to deliver high linearity in cDNA synthesis across a broad range of input RNA template for two-step quantitative RT-PCR assays.

Features of SuperScript VILO Master Mix include:
• Starting material can range up to 2.5 μg of total RNA in a 20 μL cDNA synthesis reaction
• Recombinant ribonuclease inhibitor safeguards against the degradation of target RNA due to ribonuclease contamination
• RT is engineered with reduced RNase H activity and increased thermal stability

For superior cDNA synthesis performance in RT-qPCR applications
SuperScript IV VILO Master Mix (Cat. No. 11756050) elevates the trusted VILO technology to the next level with the highly processive and thermostable SuperScript IV RT and further optimized buffer. The master mix enables efficient cDNA synthesis at higher temperatures in less time and provides superior cDNA yield and sensitivity even with suboptimal purity or scarce templates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• SuperScript VILO Master Mix (1 mL)

Store at –5 to –30°C.

Master Mix
60 min.
42°C
SuperScript III
Dry Ice
5X
250 Reactions
Reverse Transcription
RNA
qPCR
High
Reverse Transcription
250 reactions
Reduced
Real Time PCR (qPCR)
First-Strand cDNA
Master Mix
Up to 10 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2783544 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number11755250
Lot Number2486654 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number11755250
Lot Number2919509 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number11755250
Lot Number3012796 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number11755250
Lot Number2963962 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number11755250
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SuperScript™ VILO™ Master Mix > 250 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.