missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

Brand:  Thermo Scientific™ 34578

 View more versions of this product

Product Code. 15669364

  • 480.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram to high femtogram-level protein detection by western blot analysis.

SuperSignal West Pico PLUS Chemiluminescent Substrate characteristics include:

  • Sensitivity: pictogram to femtogram sensitivity
  • Stability: 8-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 6 to 24 hours
  • Recommended primary antibody concentration: 1:1000–1:5000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)

SuperSignal West Pico PLUS substrate is designed to provide excellent signal intensity and sensitivity for western blotting with HRP conjugates. The intensity of the light emission combined with the exceptional duration allows for the acquisition of multiple exposures to more easily obtain publication-quality blot images. SuperSignal West Pico PLUS substrate is compatible with different membranes, blocking reagents, and a wide range of antibody dilutions, making it an ideal choice for most western blotting applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
1 L
Horseradish Peroxidase
Picogram to high femtogram-level
120 Mini-Gel Blots, 10000 cm2 of Membrane
Sufficient for 120 mini-gel blots; 10,000 cm2 of membrane
• SuperSignal West Pico PLUS Luminol/Enhancer, 500 mL
• SuperSignal West Pico PLUS Stable Peroxide, 500 mL

Store at room temperature.
Chemiluminescence
Western Blot
SuperSignal™
HRP
2°: 1:20-100 K (10-50 ng/mL), 1°: 1:1 K (0.2-1 μg/mL)
6 to 24 hr.
SuperSignal West Pico PLUS Luminol/Enhancer, SuperSignal West Pico PLUS Stable Peroxide
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.