missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Supor™ Membrane, Gridded

Supor™ Membrane designed for environmental monitoring applications.

Brand:  Cytiva (formerly Pall Lab) 60043

338.20 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 17154481

  • 356.00 € / Pack of 200
Estimated Shipment: 30-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
PES
Sterile
200/Pk.
Bulk with Parchment Separators
0.45 μm
Black
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers