Learn More
OHAUS™ Support Stand
OHAUS™ Support Stand provides secure support for holding glassware at elevated positions.
Brand:  OHAUS™ 30400032
378.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15742449
  • 390.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Support Stand with Rod
Clamp, Support, Stand/Rod, CLR-STRODS091
267 mm
267.00 mm
17.75 in.
Stainless Steel
16 mm
10.50 in.
914 mm
451.00 mm