Learn More
OHAUS™ Support Stand Rod
OHAUS™ Support Stand Rod provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30400036
This item is not returnable.
View return policy
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Rod | |
Stainless Steel | |
Clamp, Support, Rod 46 cm, CLR-SPRODS046 | |
460 mm |