Learn More
Hellma™ Suprasil Semi Micro Flow Through Cell
Brand:  Hellma™ 1767001510-40
Product Code. 10743091
  • 733.00 € / Each
Estimated Shipment: 22-08-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Semi-micro Flow Through Cell
Suprasil
35 mm
390 μL
200 to 2500 nm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hellma™ Suprasil Semi Micro Flow Through Cell > Wavelength Range: 200 to 2500 nm