missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Stacking Buffer

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

33.40 € - 76.00 €

Specifications

Product Line SureCast™
Shipping Condition Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15558055
View Documents
Invitrogen™
HC2112
2 x 500 mL
33.40 €
Pack of 2
In Stock
to see stock.
Add to basket
 
15627968
View Documents
Invitrogen™
HC2115
5 x 500 mL
76.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank
Specifications

Specifications

SureCast™
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.