missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureLock™ Tandem Midi Blot Module

The Invitrogen SureLock Tandem Midi Blot Module is a wet transfer device used exclusively with the SureLock Tandem Midi Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total.

Brand:  Invitrogen™ STM2001

Product Code. 16854103

  • 564.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The SureLock Tandem Midi Blot Module is compatible with all Invitrogen midi gels (blot size: 8.5 cm x 14 cm). The blot module accommodates one midi gel per wet transfer. The unique design make the module easy to use, while the efficient size of the inner core uses less transfer buffer per transfer (only ∼300 mL per transfer). Since transfer buffer contains methanol, this also results in less hazardous waste requiring special disposal. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes in 30 minutes and at room temperature.

Features:

  • Integrative design—used with the SureLock Tandem Midi Gel Tank, eliminates the need for a separate transfer tank
  • Efficient size—transfer buffer needed is about one-third of traditional wet-transfer systems reducing methanol/transfer buffer waste and cost
  • Quick set-up (time & materials)
  • Compatible—efficiently transfers all Invitrogen pre-cast midi gels
  • Smart design—anode and cathode halves are hinged and connected by a slider. The design enables all parts to be permanently connected eliminating the need for multiple accessories.
  • Low voltage transfer (25 volts)—highly conductive anode plate separated by only 0.5 inches from the cathode plate enables efficient transfer at 25 volts within 30 minutes
  • Room temperature transfer—eliminates the need to pre-chill buffers overnight and the hassle and messiness of ice baths used with traditional wet-transfer systems that require 100 volts for transfer and generate excessive heat
  • Fast and efficient—30 minute transfer without compromising performance
  • Simple clean up—empty, rinse, and dry (no spills or ice disposal)
  • Optional SureLock Tandem Transfer Tray — can be used to set up wet transfer of midi gels using SureLock Tandem Blot Module
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
1 SureLock Tandem Midi Blot Module
• 1 Blotting Tweezer
• 1 Blotting Roller
• 4 Blotting Sponges
SureLock™ Tandem Midi Gel Tank
Midi (8 x 13 cm)
Wet
Blot Module
Midi/Wide Format (8 cm x 13 cm)
Room Temperature
8 x 12 x 3
Wet Transfer
Midi Precast Protein Gels
Nitrocellulose, PVDF
SureLock
SureLock Tandem Midi Gel Tank (STM1001)
1 module
1 year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.