missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE

Choose from a comprehensive line of pipet tips for demanding liquid-handling applications

97.35 € - 136.00 €

Specifications

Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Format Hinged Rack
For Use With (Equipment) Universal Fit
View More Specs

Products 8
Product Code Brand Volume (Metric) Tip Style Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Tip Style Color Price Quantity & Availability  
11903466
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TFA104-10-FIS
0.1 to 10 μL Micro Point Natural
97.35 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11913466
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TF102-10-FIS
0.1 to 10 μL Micro Point, Extended Length Natural
133.00 €
Pack of 960
In Stock
to see stock.
Add to basket
 
11977714
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TF102-20-FIS
0.1 to 20 μL Extended Length, Micro Point, Low Retention Clear
136.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11943466
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TF113-20-FIS
2 to 20 μL Beveled Natural
103.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11953466
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TF113-100-FIS
10 to 100 μL Beveled Natural
127.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11963466
GREENER_CHOICE
View Documents
Fisherbrand™
TF140-200-FIS
20 to 200 μL Micro Point Natural
107.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11987714
GREENER_CHOICE
View accessory products View Documents
Fisherbrand™
TF140-300-FIS
30 to 300 μL Micro Point Clear
135.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
11973466
GREENER_CHOICE
View Documents
Fisherbrand™
TF112-1000-FIS
100 to 1000 μL Micro Point Natural
135.00 €
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Choose from a comprehensive line of pipet tips for demanding liquid-handling applications

Universal fit for research grade pipetters.

  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable

Specifications

Specifications

Filtered
Not Autoclavable
Universal Fit
Sterile
Hinged Rack
SureOne
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.