Learn More
Thermo Scientific™ SureQuant™ AKT Pathway (Phospho) Relative Quantitation Module
Thermo Scientific™ SureQuant™ AKT Pathway (Phospho) Relative Quantitation Module provides a complete quantitation solution for the analysis of key phosphorylation events of AKT signaling pathway proteins using mass spectrometry (MS).
Brand:  Thermo Scientific™ A40087
Product Code. 17183159
  • 818.00 € / 10 reactions
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complete - contains validated AQUA HeavyPeptide mix, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
  • Validated - peptides rigorously tested for successful quantitation of each target peptide
  • Multiplex - able to quantitate phosphorylated AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 in a single reaction
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mass Spectrometry
Approved for shipment on Wet or Dry Ice
SureQuant™
Mass Spectrometry
Peptides
10 Reactions
Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Target Selection
10 reactions kit
Quantitation Module
Protein Samples