Learn More
Thermo Scientific™ SureSTART™ 11 mm Snap Caps, Level 2 High-throughput Applications
Use Level 2 Thermo Scientific™ SureSTART™ 11 mm Snap Caps with snap vials that have an 11 mm opening for your high-throughput applications, when robustness and reproducibility are key.
18.42 € - 1296.93 €
Specifications
Material Polyethylene
Level Level 2
Product Line SureSTART Level 2
Certifications/Compliance Specification Certified
Type Snap Cap
View More Specs
Products 20
Product Code Brand Color Septum Hardness Quantity Price Quantity & Availability  
Product Code Brand Color Septum Hardness Quantity Price Quantity & Availability  
17333843
View Documents
Thermo Scientific™
6ARC11NRT1
Blue Natural Rubber/Clear TEF 60 Shore 100 Pk.
23.05 €
Pack of 100
In Stock
to see stock.
Add to basket
17993601
View Documents
Thermo Scientific™
6ARC11SPI1
Blue White Silicone/Red Polyimide 45 Shore 100 Pk.
36.18 €
Pack of 100
In Stock
to see stock.
Add to basket
17343843
View Documents
Thermo Scientific™
6ARC11NRT1F
Blue Natural Rubber/Clear TEF 60 Shore 5000/Pk.
1037.55 €
Pack of 5000
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17353843
View Documents
Thermo Scientific™
6ARC11NRT1R
Red Natural Rubber/Clear TEF 60 Shore 100 Pk.
22.99 €
Pack of 100
In Stock
to see stock.
Add to basket
17363843
View Documents
Thermo Scientific™
6ARC11NRT1RF
Red Natural Rubber/Clear TEF 60 Shore 5000/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17373843
View Documents
Thermo Scientific™
6ARC11NRT1RT
Red Natural Rubber/Clear TEF 60 Shore 1000/Pk.
218.43 €
Pack of 1000
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17383843
View Documents
Thermo Scientific™
6ARC11NRT1T
Blue Natural Rubber/Clear TEF 60 Shore 1000/Pk.
218.43 €
Pack of 1000
In Stock
to see stock.
Aggiungi al carrello
17393843
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1
Blue White Silicone/Red PTFE 45 Shore 100 Pk.
28.74 €
Pack of 100
In Stock
to see stock.
Aggiungi al carrello
17303853
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1F
Blue White Silicone/Red PTFE 45 Shore 5000/Pk.
1296.93 €
Pack of 5000
Estimated Shipment: 09-04-2025
to see stock.
Aggiungi al carrello
17313853
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1R
Red White Silicone/Red PTFE 55 Shore 100 Pk.
28.74 €
Pack of 100
In Stock
to see stock.
Aggiungi al carrello
17323853
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1RF
Red White Silicone/Red PTFE 55 Shore 5000/Pk.
1296.93 €
Pack of 5000
Estimated Shipment: 23-05-2025
to see stock.
Aggiungi al carrello
17333853
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1RT
Red White Silicone/Red PTFE 55 Shore 1000/Pk.
273.05 €
Pack of 1000
Estimated Shipment: 09-04-2025
to see stock.
Aggiungi al carrello
17343853
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1T
Blue White Silicone/Red PTFE 45 Shore 1000/Pk.
273.05 €
Pack of 1000
Estimated Shipment: 09-04-2025
to see stock.
Aggiungi al carrello
17885174
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11RRT1C
Clear Red Silicone/Clear PTFE 60 Shore 100 Pk.
22.31 €
Pack of 100
In Stock
to see stock.
Aggiungi al carrello
17845184
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1C
Clear White Silicone/Red PTFE 45 Shore 100 Pk.
28.74 €
Pack of 100
In Stock
to see stock.
Add to basket
17815174
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1B
Black White Silicone/Red PTFE 45 Shore 100 Pk.
28.74 €
Pack of 100
In Stock
to see stock.
Add to basket
17835214
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1OR
Orange White Silicone/Red PTFE 45 Shore 100 Pk.
28.74 €
Pack of 100
In Stock
to see stock.
Add to basket
17875214
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11ST1ORT
Orange White Silicone/Red PTFE 45 Shore 1000/Pk.
287.41 €
Pack of 1000
In Stock
to see stock.
Add to basket
17825214
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11-P
Clear White Virgin PTFE 53 Shore 100 Pk.
18.42 €
Pack of 100
In Stock
to see stock.
Add to basket
17835174
missing translation for 'viewDocuments'
Thermo Scientific™
6ARC11RRT1
Blue Red Silicone/Clear PTFE 60 Shore 100 Pk.
22.40 €
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Use Thermo Scientific™ SureSTART™ 11 mm Snap Caps with snap vials that have an 11 mm opening, including our SureSTART 2 mL Glass Snap Vials (Level 2). Select Level 2 products for high-throughput applications, when robustness and reproducibility are key. Choose from soft polyethylene caps with double snap functionality that guarantees a tight seal. Our snap caps include silicone/PTFE or rubber/TEF septa of various hardness values designed to reduce autosampler needle issues.
Our performance Level 2 caps are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
The SureSTART Performance Levels
SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
• Level 1: Everyday analysis
Chromatography approved
SureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
• Level 2: High-throughput analysis
When robustness and reproducibility are key
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
• Level 3: High performance
When sensitivity is a must
SureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.
All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.
Specifications
Specifications
Polyethylene
SureSTART Level 2
Snap Cap
Level 2
Specification Certified
11 mm