missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Deep Well Plates, Square V-Bottom, Level 2 High-Throughput Applications

For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 96-well microplates.

77.28 € - 537.68 €

Specifications

Certified Certified, GC/MS and LC/MS Tested
Array 8 x 12
Product Line SureSTART WebSeal Level 2
Color Clear
Material Polypropylene
View More Specs

Products 2
Product Code Brand Total Volume (Metric) Well No. per Pack Price Quantity & Availability  
Product Code Brand Total Volume (Metric) Well No. per Pack Price Quantity & Availability  
15182729
View Documents
Thermo Scientific™
60180-P212
2000 μL 5 Plates
77.28 €
Pack of 5
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
15172729
View Documents
Thermo Scientific™
60180-P202
2000 μL 50 Plates
537.68 €
Pack of 50
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ SureSTART™ Level 2 WebSeal™ 96-Well Square V-Bottom Deep Well Plates are designed to ensure high quality data with an uninterrupted workflow for high-throughput GC and HPLC/UHPLC applications. They are ideal for regulated labs that use water and polar solvents.

Use Level 2 polypropylene well plates for routine QA/QC workflows, sensitive applications with lower sample concentrations, and data collection for total ion chromatograms (TIC).

These 96-well plates are certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards. Seal these well plates with 96-SureSTART WebSeal Well Plate Sealing Mats, 8 mm Diameter, Square, Level 1 or SureSTART WebSeal 96-Well Plate Sealing Mats, 8 mm Diameter, Square, Level 2.

Performance Level 2 Plates – for High Throughput Applications

  • Ideal for non-regulated labs that use water and polar solvents
  • Certified plastic well plates, seals, tapes
  • GC-MS and LC-MS tested
  • Suitable for HPLC/UHPLC applications
  • Suitable for QA/QC workflows and sensitive applications with low sample concentrations
  • Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
  • Specification Certified
  • All Thermo Scientific SureSTART WebSeal well plates are dimensionally verified

Lot Traceable

  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request

About Thermo Scientific WebSeal Well Plates & Mats

Thermo Scientific WebSeal plates and mats are available in three performance levels:

  • Level 1 well plates are for everyday applications and are suitable for non-regulated labs using water and polar solvents
  • Level 2 well plates are for high-throughput applications and are suitable for regulated labs using water and polar solvents
  • Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents
Specifications

Specifications

Certified, GC/MS and LC/MS Tested
SureSTART WebSeal Level 2
Polypropylene
50 to 1900 μL
Non-coated
Square, Conical Bottom (V-well)
44.4 mm
Deep Well
8 x 12
Clear
96
Non-sterile
2000 μL
-80°C to 121°C
7 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.