Learn More
Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Level 1 Everyday Analysis
Use SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Performance Level 1 for everyday chromatography analysis. They are specification certified and conform to ANSI-SLAS 4-2004 standards.
55.53 € - 285.42 €
Specifications
Sterility Non-sterile
No. of Wells 96
Plug Diameter 8 mm
For Use With (Application) HPLC, UHPLC, GC
Product Line SureSTART WebSeal Level 1
View More Specs
Products 8
Product Code Brand Pre-slit Material Solvent Or Alcohol Resistant No. per Pack Price Quantity & Availability  
Product Code Brand Pre-slit Material Solvent Or Alcohol Resistant No. per Pack Price Quantity & Availability  
15192709
View Documents
Thermo Scientific™
60180-M101
No EVA Alcohol Resistant 5 Mats
55.53 €
Pack of 5
Estimated Shipment: 14-05-2025
Add to basket
17443193
View Documents
Thermo Scientific™
60180-M171
No EVA Alcohol Resistant
197.99 €
Pack of 50
17433193
View Documents
Thermo Scientific™
60180-M134
No Silicone No Resistance 5 Mats
71.61 €
Pack of 5
15896251
View Documents
Thermo Scientific™
60180-M184
No Silicone No Resistance 5 Mats
285.42 €
Pack of 50
In Stock
Add to basket
16231362
View Documents
Thermo Scientific™
60180-M183
Yes Silicone No Resistance 5 Mats
86.93 €
Pack of 5
Estimated Shipment: 16-05-2025
Add to basket
15182709
View Documents
Thermo Scientific™
60180-M100
Yes Silicone No Resistance 10 Mats
129.65 €
Pack of 10
Estimated Shipment: 09-05-2025
Add to basket
15102719
View Documents
Thermo Scientific™
60180-M102
No Silicone, PTFE Solvent Resistant 5 Mats
70.05 €
Pack of 5
Estimated Shipment: 09-05-2025
Add to basket
15112719
View Documents
Thermo Scientific™
60180-M103
Yes Silicone, PTFE Solvent Resistant 5 Mats
87.89 €
Pack of 5
In Stock
Add to basket
Description
Show More
Specifications
Specifications
Non-sterile
8 mm
SureSTART WebSeal Level 1
Yes
WebSeal 96-well Round Well Plate
96
HPLC, UHPLC, GC
None
Round
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ SureSTART™ WebSeal™ 96-Well Plate Sealing Mats, 8 mm Diameter, Round, Level 1 Everyday Analysis