Learn More
3M™ Clean-Trace™ Surface Protein (Allergen) Test Swab ALLTEC60
Rapid, simple, and reliable solution to monitor the presence of allergens on surfaces to effectively manage cleaning.
Brand:  3M™ Clean-Trace™ 7000034528
Encompass
Product Code. 17228530
  • 299.78 € / 60 tests
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detects as little as 3 μg of protein on surfaces and in hard to reach areas.
  • Provides a tool to help assess and improve hygienic performance.
  • Visual reading of a color change indicates the level of cleanliness.
  • Real time results allow immediate corrective action.
  • Validated for a range of allergenic proteins, including egg, milk, gluten, soy, peanut, almond and buckwheat.
Specifications
Specifications
Surface Hygiene
60/Cs.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 3M Clean-Trace Surface Protein (Allergen) Test Swab
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : copper sulphate pentahydrate, sodium hydroxide