Learn More
Novus Biologicals™ Surfactant Protein A Antibody Pair
Brand:  Novus Biologicals™ H00653509-AP41
Product Code. 18563394
  • 1100.00 € / 1 set
Explore more special offers
Specifications
3 ng/mL to 100 ng/mL
Sandwich ELISA
653509
1 Set
COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B
1 x PBS, pH 7.4
Unconjugated
Sandwich ELISA
SFTPA1
Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles.