Learn More
BICASA Suspended Undercounter Cabinet with Drawers for Kompatto Workstation
Suspended Undercounter cabinet with 3 soft-close drawers with central lock for Kompatto workstation. Made in Fire-retardant particleboard.
Brand:  BICASA K-DRAW-R
 View more versions of this product
Product Code. 30057560
  • 711.00 € / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Kompatto Series Workstation
  • Suspended under the countertop
  • Drawers: EN 14749 certified sliding guide - Soft-Close damping system - Single loading capacity of Kg 40,0
  • Central locking system as standard
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
  • Fire-retardant particleboard body
  • Light Gray color (2 decorative) - mm 19 thickness
  • Frontal drawer panels and doors with ABS rounded rim
Specifications
Specifications
Suspended Undercounter Cabinet
Red
3 Soft-close Drawers with Central Lock
600 x (500 + 20) x 530 mm
500 mm
Light Gray
Particleboard
Kompatto Series Workstation
19.7 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BICASA Suspended Undercounter Cabinet with Drawers for Kompatto Workstation > Includes: 3 Soft-close Drawers with Central Lock, Color Facade: Red