Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tubes, Sustain™ Series
GREENER_CHOICE
Product Code. p-10504677 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
0.6 mL
1.5 mL
2 mL
Unit Size:
Pack of 1000
Pack of 500
Product Code. 30130202
Brand: Thermo Scientific™ 3400DLB
Only null left
Add to basket
Edge
Add to basket
The Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tubes, Sustain Series offer a greener solution for laboratory researchers in biotech, pharma, and forensics. The tubes are made from biobased polypropylene allocated on a mass balance basis derived from second-generation waste and residue oils.
We are committed to designing our products with the environment in mind. The DNA Low Binding Snap Cap Microcentrifuge Tubes, Sustain Series, are ideal for general lab researchers in biotech, pharma, and forensics who need to ensure nucleic acid integrity in precious samples and are looking for more sustainable products.
DNA integrity is critical for any genomics workflow, and scientists rely on products to help reduce loss of sample DNA. Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tubes are designed without the use of per- and polyfluoroalkyl substances (PFAS) to help minimize DNA sample loss.
The polypropylene used in the DNA Low Binding Snap Cap Microcentrifuge Tubes, Sustain Series, consists of ISCC PLUS–certified, second-generation, biobased material.
By choosing these more sustainable tubes, customers can reduce their carbon footprint, support their Scope 3 emission reduction targets, and contribute to a circular economy, all while maintaining the same performance standards as fossil fuel–based alternatives.
The material reduces carbon dioxide equivalents (CO2 e) by 3.43 kg per kg of biobased polypropylene, relative to the non–Sustain Series equivalent.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Natural
Material Polypropylene Resin
Volume (Metric) 0.6 mL
Sterility Non-sterile
Graduated Yes
Closure Type Snap Cap
Fit for Purpose PCR Ready
Bottom Shape Conical
Green Features Less waste, renewable material
Shape Cylindrical
Product Type Microcentrifuge Tube