missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SVL™ Borosilicate Glass Lids
For use with SVL™ reaction vessels
Brand: SVL 296-03
This item is not returnable.
View return policy
Description
- For use with SVL™ reaction vessels
- Borosilicate glass construction available in three styles: - Plain with knob (no necks) to act as a cover - 296-00 - With two necks (one vertical central neck and one vertical side neck) - With five necks as shown in the table below
Specifications
Borosilicate Glass | |
100 mm | |
100 mm |
Reaction Vessel | |
Lid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction