Learn More
Quickfit™ Claisen Still Head Adapter
PTFE joint sleeves eliminate the possibility of joint seizure
Brand: Quickfit SH6/44
This item is not returnable.
View return policy
Description
- Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer