Learn More
SVL™ Cone/Screw Thread Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
Brand:  SVL 670-70
426.55 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15577740
  • 461.00 € / Pack of 10
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/23
Borosilicate Glass
15
SVL™ and Quickfit™ Glassware
Cone/Screw thread adapter