Learn More
SVL™ Glass Screwthreads
For use by glassblowers to construct apparatus with SVR™ joints
Brand:  SVL 703-02
 View more versions of this product
Product Code. 15507750
  • 305.00 € / Pack of 50
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use by Glassblowers to construct apparatus with SVL™ joints
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
6.49 in.
1.8 mm
Junction Tube
22.5 mm
165 mm
For use by Glassblowers to construct apparatus with SVL™ joints