Learn More
SVL™ Glass Screwthreads
For use by glassblowers to construct apparatus with SVR™ joints
Brand: SVL 703-04
1472.50 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use by Glassblowers to construct apparatus with SVL™ joints
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
Specifications
Junction Tube | |
1.7 in. | |
6.49 in. | |
3.2 mm |
Borosilicate Glass (3.3) | |
44.1 mm | |
165 mm | |
For use by Glassblowers to construct apparatus with SVL™ joints |