Learn More
SVL™ Jacketed Reaction Vessel
Ideal for temperature controlled reactions.
Brand:  SVL 295-01
Product Code. 15557690
  • 9270.00 € / Pack of 5
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With outer jacket through which a temperature controlling medium can be circulated (maximum temperature of 200°C)
  • Contents of inner chamber can be run off via a Torion PTFE stopcock
  • When used with an SVL™ lid and PTFE covered sealing ring, the assembly is grease free and resistant to chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 mL
Glass
Ideal for temperature controlled reactions and for circulation of water or oil etc
Jacketed reaction vessel