Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Switch stirring heads quickly and easily. Pyrex™ SVL™ Stirrer Head for SVL™ Stirrer Shafts are constructed of durable PTFE polymer; some types are fortified with a metal core.
Brand:  Pyrex™ 588-93
 View more versions of this product
Product Code. 12024422
  • 90.30 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for use with SVL™ stirrer shafts
  • Head simply screws into the end of the shaft
  • Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
TRUSTED_SUSTAINABILITY
Specifications
Specifications
70 mm
Vessels with SVL™ 30 joint or 29/32 socket
1 Each
Collapsible Impeller